Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATCTCCAGGCTGTCTGCCTCTTTT[G/T]CCAACAAGTTCTCTTTTTCACACAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610872 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF19B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RNF19B - ring finger protein 19B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127361.2 | 3033 | Intron | NP_001120833.1 | |||
NM_001300826.1 | 3033 | Intron | NP_001287755.1 | |||
NM_153341.3 | 3033 | Intron | NP_699172.2 | |||
XM_006710356.2 | 3033 | UTR 3 | XP_006710419.1 | |||
XM_006710357.3 | 3033 | Intron | XP_006710420.1 | |||
XM_006710358.3 | 3033 | Intron | XP_006710421.1 |