Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGATGAACAAAAATCACCCTATAC[C/T]ATACTTGTGCACAATAATGTTCAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
19 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605494 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EFCAB7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EFCAB7 - EF-hand calcium binding domain 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032437.3 | Intron | NP_115813.2 | ||||
XM_005271274.3 | Intron | XP_005271331.1 | ||||
XM_006710976.3 | Intron | XP_006711039.1 | ||||
XM_006710977.1 | Intron | XP_006711040.1 | ||||
XM_011542301.2 | Intron | XP_011540603.1 | ||||
XM_017002548.1 | Intron | XP_016858037.1 |
ITGB3BP - integrin subunit beta 3 binding protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |