Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTTACCCTCTGTTGTAAGAAATA[C/T]TAGCTTTTCTCTGTGTATGTATGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 103030 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AK4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AK4 - adenylate kinase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005353.2 | Intron | NP_001005353.1 | ||||
NM_013410.3 | Intron | NP_037542.1 | ||||
NM_203464.2 | Intron | NP_982289.1 | ||||
XM_017000612.1 | Intron | XP_016856101.1 | ||||
XM_017000613.1 | Intron | XP_016856102.1 |