Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTCAGGAAAACCTCTCAACTCTG[C/T]CCCTTACTTTCCATCCTTACTCCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610133 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ST6GALNAC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ST6GALNAC3 - ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160011.1 | Intron | NP_001153483.1 | ||||
NM_152996.2 | Intron | NP_694541.2 | ||||
XM_006710553.3 | Intron | XP_006710616.1 | ||||
XM_017000937.1 | Intron | XP_016856426.1 | ||||
XM_017000938.1 | Intron | XP_016856427.1 | ||||
XM_017000939.1 | Intron | XP_016856428.1 | ||||
XM_017000940.1 | Intron | XP_016856429.1 | ||||
XM_017000941.1 | Intron | XP_016856430.1 | ||||
XM_017000942.1 | Intron | XP_016856431.1 |