Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCTGCTTCTTAGGTACAAGAGGA[A/C]GAAGAATCTGTACAGAACCAAAAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
18 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611327 MIM: 603444 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJB4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAJB4 - DnaJ heat shock protein family (Hsp40) member B4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317099.1 | Intron | NP_001304028.1 | ||||
NM_001317100.1 | Intron | NP_001304029.1 | ||||
NM_001317101.1 | Intron | NP_001304030.1 | ||||
NM_001317102.1 | Intron | NP_001304031.1 | ||||
NM_001317103.1 | Intron | NP_001304032.1 | ||||
NM_007034.4 | Intron | NP_008965.2 |
FUBP1 - far upstream element binding protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |