Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCATTCTACTGTTCACTGCTATC[C/T]TCCTCCAAACTTTACTCTGCACTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
24 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610468 MIM: 613975 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IFI44 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IFI44 - interferon induced protein 44 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006417.4 | 74 | Intron | NP_006408.3 | |||
XM_005270380.3 | 74 | UTR 5 | XP_005270437.1 | |||
XM_011540516.2 | 74 | UTR 5 | XP_011538818.1 | |||
XM_017000083.1 | 74 | UTR 5 | XP_016855572.1 |
IFI44L - interferon induced protein 44 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |