Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAATAGTGTTAGAAACAGATCTGG[G/A]TGCTGGGTATGCTCATTGCTACTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608848 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FNBP1L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FNBP1L - formin binding protein 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024948.2 | Intron | NP_001020119.1 | ||||
NM_001164473.2 | Intron | NP_001157945.1 | ||||
NM_017737.4 | Intron | NP_060207.2 | ||||
XM_011541625.2 | Intron | XP_011539927.1 | ||||
XM_017001533.1 | Intron | XP_016857022.1 |