Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCTGGTGGCCCCGGCTGAGGCTG[G/T]TATTTCCGCAGGTAGCGCTGCGCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 609437 | ||||||||||||||||||||
Literature Links: |
STK40 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
STK40 - serine/threonine kinase 40 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282546.1 | 1758 | Silent Mutation | NP_001269475.1 | |||
NM_001282547.1 | 1758 | Silent Mutation | NP_001269476.1 | |||
NM_032017.2 | 1758 | Silent Mutation | NP_114406.1 |