Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAATGACATGGCTCTACCTAACTG[C/T]AGGCTGAAAGAAAAGGAACAAGGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
24 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 248610 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DBT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DBT - dihydrolipoamide branched chain transacylase E2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001918.3 | 7320 | UTR 3 | NP_001909.3 | |||
XM_005270545.3 | 7320 | Intron | XP_005270602.1 | |||
XM_017000468.1 | 7320 | Intron | XP_016855957.1 | |||
XM_017000469.1 | 7320 | Intron | XP_016855958.1 |