Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATGAAGCGTATATTTGAGCCCAA[C/G]TGCTTGGACGCCTTCCTAAACTTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
22 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138385 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GSTM5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GSTM5 - glutathione S-transferase mu 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000851.3 | 616 | Missense Mutation | AAC,AAG | N,K 173 | NP_000842.2 | |
XM_005270784.4 | 616 | Missense Mutation | AAC,AAG | N,K 173 | XP_005270841.1 | |
XM_005270785.4 | 616 | Missense Mutation | AAC,AAG | N,K 69 | XP_005270842.1 |