Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAATGGGGTCCACTCACACCTGCA[C/T]CCCCTGGCCCTGCAGCTGGAGCACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
25 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609977 MIM: 611123 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDCA8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDCA8 - cell division cycle associated 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EPHA10 - EPH receptor A10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099439.1 | 3102 | Missense Mutation | ATG,GTG | M,V 1006 | NP_001092909.1 | |
NM_173641.2 | 3102 | Intron | NP_775912.2 | |||
XM_017001080.1 | 3102 | Missense Mutation | ATG,GTG | M,V 609 | XP_016856569.1 | |
XM_017001081.1 | 3102 | UTR 3 | XP_016856570.1 |