Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTATGTGTGGAAGGAGCACAGGATT[A/G]GTGTGAAAGGCTTCTAGGTGGCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
20 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 162361 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NHLH2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NHLH2 - nescient helix-loop-helix 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001111061.1 | Intron | NP_001104531.1 | ||||
NM_005599.3 | Intron | NP_005590.1 | ||||
XM_017001370.1 | Intron | XP_016856859.1 |