Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTGATTTTTATCTCTTTAGATGT[A/G]TTTTGATTGTTCTTGAACGTTATAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604718 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101929099 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101929099 - uncharacterized LOC101929099 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TTF2 - transcription termination factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003594.3 | Intron | NP_003585.3 | ||||
XM_005271277.3 | Intron | XP_005271334.1 | ||||
XM_011542303.2 | Intron | XP_011540605.1 | ||||
XM_017002549.1 | Intron | XP_016858038.1 | ||||
XM_017002550.1 | Intron | XP_016858039.1 | ||||
XM_017002551.1 | Intron | XP_016858040.1 | ||||
XM_017002552.1 | Intron | XP_016858041.1 | ||||
XM_017002553.1 | Intron | XP_016858042.1 | ||||
XM_017002554.1 | Intron | XP_016858043.1 | ||||
XM_017002555.1 | Intron | XP_016858044.1 |