Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCTTGGGTATATGTCTTCGGCCA[A/G]AAAAAAAAAAAAAAAAAAAACACGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 604127 | ||||||||||||||||||||
Literature Links: |
TBX15 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TBX15 - T-box 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152380.2 | 2389 | UTR 3 | NP_689593.2 | |||
XM_005271161.3 | 2389 | UTR 3 | XP_005271218.1 | |||
XM_005271162.1 | 2389 | UTR 3 | XP_005271219.1 | |||
XM_011542052.2 | 2389 | UTR 3 | XP_011540354.1 |