Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTACTGCTTGTTTCATTTACTTCA[C/T]AGAATTGGGAAATAAATGAGATAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608973 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SIK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SIK2 - salt inducible kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015191.2 | Intron | NP_056006.1 | ||||
XM_017017417.1 | Intron | XP_016872906.1 | ||||
XM_017017418.1 | Intron | XP_016872907.1 |