Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTGGACGTCCAGGATGGAGTGAG[C/T]TGCACACCCCTGCAACTGGCCCTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608774 MIM: 126450 | ||||||||||||||||||||
Literature Links: |
ANKK1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANKK1 - ankyrin repeat and kinase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178510.1 | 4337 | Silent Mutation | AGC,AGT | S,S 725 | NP_848605.1 | |
XM_011542736.2 | 4337 | Silent Mutation | AGC,AGT | S,S 736 | XP_011541038.1 | |
XM_011542737.2 | 4337 | Silent Mutation | AGC,AGT | S,S 726 | XP_011541039.1 | |
XM_011542738.2 | 4337 | Silent Mutation | AGC,AGT | S,S 662 | XP_011541040.1 | |
XM_017017475.1 | 4337 | Silent Mutation | AGC,AGT | S,S 735 | XP_016872964.1 |
DRD2 - dopamine receptor D2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |