Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCCCAAGCGCTGGCAGTGACGGG[A/G]ACAAAGGTCTAAAGCCCAGGGAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601814 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FXYD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FXYD2 - FXYD domain containing ion transport regulator 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001680.4 | 985 | UTR 3 | NP_001671.2 | |||
NM_021603.3 | 985 | UTR 3 | NP_067614.1 |
FXYD6-FXYD2 - FXYD6-FXYD2 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204268.1 | 985 | UTR 3 | NP_001191197.1 | |||
NM_001243598.2 | 985 | UTR 3 | NP_001230527.1 |