Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCGGGGTGCTGGGAGCACTGCAG[A/G]GTCCACCCTGACTGGGGCACAGAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606565 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMPRSS4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMPRSS4 - transmembrane protease, serine 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083947.1 | Intron | NP_001077416.1 | ||||
NM_001173551.1 | Intron | NP_001167022.1 | ||||
NM_001173552.1 | Intron | NP_001167023.1 | ||||
NM_001290094.1 | Intron | NP_001277023.1 | ||||
NM_001290096.1 | Intron | NP_001277025.1 | ||||
NM_019894.3 | Intron | NP_063947.1 | ||||
XM_005271613.3 | Intron | XP_005271670.1 | ||||
XM_005271614.2 | Intron | XP_005271671.1 | ||||
XM_005271615.3 | Intron | XP_005271672.1 | ||||
XM_011542901.2 | Intron | XP_011541203.1 | ||||
XM_011542902.2 | Intron | XP_011541204.1 | ||||
XM_011542903.2 | Intron | XP_011541205.1 | ||||
XM_011542904.2 | Intron | XP_011541206.1 |
TMPRSS4-AS1 - TMPRSS4 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |