Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGACCTGACCTGTCCTTCCTCTCT[A/C]CCCCCAGATTCCCATCCAGGACTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609004 MIM: 601613 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCL9L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BCL9L - B-cell CLL/lymphoma 9-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182557.2 | 5823 | UTR 3 | NP_872363.1 | |||
XM_005271511.2 | 5823 | UTR 3 | XP_005271568.1 | |||
XM_005271512.2 | 5823 | UTR 3 | XP_005271569.1 | |||
XM_005271513.2 | 5823 | UTR 3 | XP_005271570.1 | |||
XM_006718815.2 | 5823 | UTR 3 | XP_006718878.1 | |||
XM_011542756.2 | 5823 | UTR 3 | XP_011541058.1 | |||
XM_017017572.1 | 5823 | UTR 3 | XP_016873061.1 | |||
XM_017017573.1 | 5823 | UTR 3 | XP_016873062.1 | |||
XM_017017574.1 | 5823 | UTR 3 | XP_016873063.1 |
CXCR5 - C-X-C motif chemokine receptor 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC101929133 - uncharacterized LOC101929133 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |