Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTAACCACAGAGGTACTGCAGAAG[A/G]AACCAGTGAGCTGTCCCTCCCTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601746 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HYOU1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HYOU1 - hypoxia up-regulated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130991.2 | 3268 | UTR 3 | NP_001124463.1 | |||
NM_006389.4 | 3268 | UTR 3 | NP_006380.1 | |||
XM_005271392.3 | 3268 | UTR 3 | XP_005271449.1 | |||
XM_005271393.3 | 3268 | UTR 3 | XP_005271450.1 | |||
XM_005271394.3 | 3268 | UTR 3 | XP_005271451.1 | |||
XM_017017095.1 | 3268 | UTR 3 | XP_016872584.1 | |||
XM_017017096.1 | 3268 | UTR 3 | XP_016872585.1 | |||
XM_017017097.1 | 3268 | UTR 3 | XP_016872586.1 |