Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAACGTTTAAACCTAAGGAATGGT[A/G]ATGTTTTCCAACAGTACTGCTTGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602005 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105369536 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105369536 - uncharacterized LOC105369536 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SORL1 - sortilin related receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003105.5 | Intron | NP_003096.1 | ||||
XM_011542963.2 | Intron | XP_011541265.1 | ||||
XM_011542965.2 | Intron | XP_011541267.1 | ||||
XM_011542967.2 | Intron | XP_011541269.1 | ||||
XM_017018169.1 | Intron | XP_016873658.1 | ||||
XM_017018170.1 | Intron | XP_016873659.1 | ||||
XM_017018171.1 | Intron | XP_016873660.1 | ||||
XM_017018172.1 | Intron | XP_016873661.1 |