Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGAAGACACCTTCTCTTTTTCCA[C/T]AGTAGTGCTGGAAGGGGGCTTGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
OR8D1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OR8D1 - olfactory receptor family 8 subfamily D member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OR8D2 - olfactory receptor family 8 subfamily D member 2 (gene/pseudogene) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002918.1 | 802 | Missense Mutation | ATG,GTG | M,V 268 | NP_001002918.1 |
OR8G5 - olfactory receptor family 8 subfamily G member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005198.1 | 802 | Intron | NP_001005198.1 | |||
XM_011542662.2 | 802 | Intron | XP_011540964.1 | |||
XM_011542663.2 | 802 | Intron | XP_011540965.1 | |||
XM_011542664.2 | 802 | Intron | XP_011540966.1 | |||
XM_011542665.2 | 802 | Intron | XP_011540967.1 | |||
XM_011542666.2 | 802 | Intron | XP_011540968.1 | |||
XM_011542667.2 | 802 | Intron | XP_011540969.1 |