Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCTCTAGACCGAAACCAAAGCTA[C/T]CCACAGCCACTGCCAGGGCCCGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608630 MIM: 607528 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ROBO3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ROBO3 - roundabout guidance receptor 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ROBO4 - roundabout guidance receptor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301088.1 | 2884 | Missense Mutation | GAT,GGT | D,G 770 | NP_001288017.1 | |
NM_019055.5 | 2884 | Missense Mutation | GAT,GGT | D,G 915 | NP_061928.4 | |
XM_006718861.1 | 2884 | Missense Mutation | GAT,GGT | D,G 877 | XP_006718924.1 | |
XM_011542875.1 | 2884 | Missense Mutation | GAT,GGT | D,G 473 | XP_011541177.1 |