Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTGGGTGCTTAAAGATGAACAGT[C/T]CGAGGATGCTGTTTCTATTCTGCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC15 - coiled-coil domain containing 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025004.2 | 76 | Intron | NP_079280.2 | |||
XM_017018346.1 | 76 | Intron | XP_016873835.1 | |||
XM_017018347.1 | 76 | Intron | XP_016873836.1 | |||
XM_017018348.1 | 76 | Intron | XP_016873837.1 | |||
XM_017018349.1 | 76 | UTR 5 | XP_016873838.1 | |||
XM_017018350.1 | 76 | Intron | XP_016873839.1 | |||
XM_017018351.1 | 76 | Intron | XP_016873840.1 | |||
XM_017018352.1 | 76 | Intron | XP_016873841.1 | |||
XM_017018353.1 | 76 | Intron | XP_016873842.1 |