Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATTTTCATTACATTTGCTTAGGC[C/T]GTTATTAGAGTTGTAACACATATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 104776 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
APLP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
APLP2 - amyloid beta precursor like protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142276.1 | Intron | NP_001135748.1 | ||||
NM_001142277.1 | Intron | NP_001135749.1 | ||||
NM_001142278.1 | Intron | NP_001135750.1 | ||||
NM_001243299.1 | Intron | NP_001230228.1 | ||||
NM_001642.2 | Intron | NP_001633.1 |