Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTGATGCTTAGAAATAAGGGGCT[C/T]ATCTTACTAATCCTATTGATGCCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 172470 MIM: 607940 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PHKG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PHKG1 - phosphorylase kinase catalytic subunit gamma 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258459.1 | Intron | NP_001245388.1 | ||||
NM_001258460.1 | Intron | NP_001245389.1 | ||||
NM_006213.4 | Intron | NP_006204.1 | ||||
XM_005271772.4 | Intron | XP_005271829.1 | ||||
XM_017012324.1 | Intron | XP_016867813.1 | ||||
XM_017012325.1 | Intron | XP_016867814.1 | ||||
XM_017012326.1 | Intron | XP_016867815.1 | ||||
XM_017012327.1 | Intron | XP_016867816.1 |
SUMF2 - sulfatase modifying factor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |