Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAAAGGTGTAAAGAGGTCTCCAA[C/T]AACAGCAGTGGCAAACGGGAGTTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611648 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GIN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GIN1 - gypsy retrotransposon integrase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPIP5K2 - diphosphoinositol pentakisphosphate kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276277.1 | Intron | NP_001263206.1 | ||||
NM_001281471.1 | Intron | NP_001268400.1 | ||||
NM_015216.3 | Intron | NP_056031.2 | ||||
XM_005271935.1 | Intron | XP_005271992.1 | ||||
XM_005271936.1 | Intron | XP_005271993.1 | ||||
XM_005271938.1 | Intron | XP_005271995.1 | ||||
XM_005277534.1 | Intron | XP_005277591.1 | ||||
XM_005277535.1 | Intron | XP_005277592.1 | ||||
XM_005277536.1 | Intron | XP_005277593.1 | ||||
XM_005277538.1 | Intron | XP_005277595.1 | ||||
XM_005277539.1 | Intron | XP_005277596.1 | ||||
XM_005277541.1 | Intron | XP_005277598.1 | ||||
XM_006714576.1 | Intron | XP_006714639.1 | ||||
XM_006714577.1 | Intron | XP_006714640.1 | ||||
XM_011543290.2 | Intron | XP_011541592.1 | ||||
XM_017009269.1 | Intron | XP_016864758.1 | ||||
XM_017009270.1 | Intron | XP_016864759.1 | ||||
XM_017009271.1 | Intron | XP_016864760.1 | ||||
XM_017009272.1 | Intron | XP_016864761.1 | ||||
XM_017009273.1 | Intron | XP_016864762.1 | ||||
XM_017009274.1 | Intron | XP_016864763.1 | ||||
XM_017009275.1 | Intron | XP_016864764.1 |