Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGATGGCTCTTAGAGCATTAAAAT[A/C]AATTTACCCATCCTTAGTTTTTGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA0825 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIAA0825 - KIAA0825 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145678.1 | Intron | NP_001139150.1 | ||||
NM_173665.2 | Intron | NP_775936.1 | ||||
XM_006714588.2 | Intron | XP_006714651.1 | ||||
XM_011543324.2 | Intron | XP_011541626.1 | ||||
XM_011543325.2 | Intron | XP_011541627.1 | ||||
XM_011543327.2 | Intron | XP_011541629.1 | ||||
XM_011543328.2 | Intron | XP_011541630.1 | ||||
XM_011543329.2 | Intron | XP_011541631.1 | ||||
XM_011543330.2 | Intron | XP_011541632.1 | ||||
XM_011543331.1 | Intron | XP_011541633.1 | ||||
XM_011543332.1 | Intron | XP_011541634.1 | ||||
XM_011543333.2 | Intron | XP_011541635.1 | ||||
XM_011543334.2 | Intron | XP_011541636.1 | ||||
XM_011543335.2 | Intron | XP_011541637.1 | ||||
XM_017009373.1 | Intron | XP_016864862.1 | ||||
XM_017009374.1 | Intron | XP_016864863.1 | ||||
XM_017009375.1 | Intron | XP_016864864.1 | ||||
XM_017009376.1 | Intron | XP_016864865.1 | ||||
XM_017009377.1 | Intron | XP_016864866.1 |