Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAGCACAATGATATCAATCAAGC[G/T]AAATGAATGCTGGTGTTATCACAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608767 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FEM1C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FEM1C - fem-1 homolog C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020177.2 | 2594 | UTR 3 | NP_064562.1 | |||
XM_005272035.4 | 2594 | UTR 3 | XP_005272092.1 | |||
XM_017009647.1 | 2594 | UTR 3 | XP_016865136.1 |