Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATCCAACACGAGTTGTGGTGGTT[C/T]TTTTATCTGAGAAAAAGCTTTTCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LOC101927100 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC101927100 - uncharacterized LOC101927100 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMED7 - transmembrane p24 trafficking protein 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181836.5 | 1034 | Missense Mutation | AAA,AGA | K,R 216 | NP_861974.1 |
TMED7-TICAM2 - TMED7-TICAM2 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164468.3 | 1034 | Intron | NP_001157940.1 | |||
NM_001164469.3 | 1034 | Intron | NP_001157941.1 |