Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGTGCTGCAGTTAGGAAGCTCAG[G/T]AAGCTACTATAGTATCCCAAGTAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
SNX24 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SNX24 - sorting nexin 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014035.2 | Intron | NP_054754.1 | ||||
XM_005271972.4 | Intron | XP_005272029.1 | ||||
XM_006714592.3 | Intron | XP_006714655.1 | ||||
XM_011543349.2 | Intron | XP_011541651.1 | ||||
XM_011543350.2 | Intron | XP_011541652.1 | ||||
XM_011543352.1 | Intron | XP_011541654.1 | ||||
XM_017009394.1 | Intron | XP_016864883.1 | ||||
XM_017009395.1 | Intron | XP_016864884.1 | ||||
XM_017009396.1 | Intron | XP_016864885.1 |