Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATTTTTTAATATCATTAAATGTT[C/T]CTCTAAATTGTTTTATAATATTCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613446 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP120 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CEP120 - centrosomal protein 120 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166226.1 | Intron | NP_001159698.1 | ||||
NM_153223.3 | Intron | NP_694955.2 | ||||
XM_005271901.4 | Intron | XP_005271958.1 | ||||
XM_011543185.2 | Intron | XP_011541487.1 | ||||
XM_011543186.2 | Intron | XP_011541488.1 | ||||
XM_017009085.1 | Intron | XP_016864574.1 |