Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTTACACATACTTACAAAATTTG[A/G]AGACAAGTTATGCTTATAAAGCTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610594 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FNIP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FNIP1 - folliculin interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008738.2 | 3356 | Missense Mutation | CCA,TCA | P,S 1072 | NP_001008738.2 | |
NM_133372.2 | 3356 | Missense Mutation | CCA,TCA | P,S 1100 | NP_588613.2 |