Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTCATTACTTTCAAGAATAACTA[C/T]AATATTGCTAGACAGTTCATTACAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604443 MIM: 616232 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACSL6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACSL6 - acyl-CoA synthetase long-chain family member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009185.2 | 2088 | UTR 3 | NP_001009185.1 | |||
NM_001205247.1 | 2088 | UTR 3 | NP_001192176.1 | |||
NM_001205248.1 | 2088 | UTR 3 | NP_001192177.1 | |||
NM_001205250.1 | 2088 | UTR 3 | NP_001192179.1 | |||
NM_001205251.1 | 2088 | UTR 3 | NP_001192180.1 | |||
NM_015256.3 | 2088 | UTR 3 | NP_056071.2 |
MEIKIN - meiotic kinetochore factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |