Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCATTTAATGCATGTATTTGTCCA[C/T]GTTTATGGCTACCATTTTCTTAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604683 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIF3A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIF3A - kinesin family member 3A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300791.1 | Intron | NP_001287720.1 | ||||
NM_001300792.1 | Intron | NP_001287721.1 | ||||
NM_007054.6 | Intron | NP_008985.3 | ||||
XM_006714526.3 | Intron | XP_006714589.1 | ||||
XM_017008995.1 | Intron | XP_016864484.1 | ||||
XM_017008996.1 | Intron | XP_016864485.1 |