Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGTTAATGCCAATTGTTATGTGCT[A/G]GGTTCTCTAACTAGGTACTATTATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605604 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAIP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PAIP2 - poly(A) binding protein interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033112.2 | Intron | NP_001028284.1 | ||||
NM_016480.4 | Intron | NP_057564.3 | ||||
XM_011543428.1 | Intron | XP_011541730.1 | ||||
XM_017009522.1 | Intron | XP_016865011.1 | ||||
XM_017009523.1 | Intron | XP_016865012.1 |