Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTGAGTCTGAGTGACACATCTGA[A/G]TTTCCACTCCTGAGTGAGGAAGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607423 MIM: 609328 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
POMT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
POMT1 - protein O-mannosyltransferase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UCK1 - uridine-cytidine kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135954.2 | 1047 | UTR 3 | NP_001129426.1 | |||
NM_001261450.2 | 1047 | UTR 3 | NP_001248379.1 | |||
NM_001261451.2 | 1047 | UTR 3 | NP_001248380.1 | |||
NM_001318519.1 | 1047 | UTR 3 | NP_001305448.1 | |||
NM_031432.3 | 1047 | UTR 3 | NP_113620.1 | |||
XM_005272224.2 | 1047 | UTR 3 | XP_005272281.1 |