Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTAGCTAGGGAACTATGTGCATAC[C/T]GAAACAGGTAAATGAACTCATCACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608465 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SETX PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SETX - senataxin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015046.5 | 3679 | UTR 3 | NP_055861.3 | |||
XM_005272171.1 | 3679 | UTR 3 | XP_005272228.1 | |||
XM_005272172.2 | 3679 | Intron | XP_005272229.1 | |||
XM_005272173.2 | 3679 | Intron | XP_005272230.1 | |||
XM_011518404.2 | 3679 | Intron | XP_011516706.1 | |||
XM_011518405.2 | 3679 | Intron | XP_011516707.1 | |||
XM_011518406.2 | 3679 | Intron | XP_011516708.1 | |||
XM_011518407.1 | 3679 | Intron | XP_011516709.1 | |||
XM_011518408.2 | 3679 | Intron | XP_011516710.1 | |||
XM_017014495.1 | 3679 | UTR 3 | XP_016869984.1 | |||
XM_017014496.1 | 3679 | UTR 3 | XP_016869985.1 | |||
XM_017014497.1 | 3679 | UTR 3 | XP_016869986.1 |