Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGGTTGAGCGCCTGCAGGCCCCC[A/G]AAGCCCCCCGGGGGCAGCTCCACGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612277 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAMTSL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADAMTSL2 - ADAMTS like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FAM163B - family with sequence similarity 163 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080515.2 | 762 | Silent Mutation | TTC,TTT | F,F 138 | NP_001073984.1 | |
XM_005272203.4 | 762 | Silent Mutation | TTC,TTT | F,F 138 | XP_005272260.1 | |
XM_005272204.4 | 762 | Silent Mutation | TTC,TTT | F,F 138 | XP_005272261.1 | |
XM_006717233.3 | 762 | Silent Mutation | TTC,TTT | F,F 138 | XP_006717296.1 | |
XM_011518912.2 | 762 | Silent Mutation | TTC,TTT | F,F 138 | XP_011517214.1 | |
XM_011518913.2 | 762 | Silent Mutation | TTC,TTT | F,F 138 | XP_011517215.1 | |
XM_017015017.1 | 762 | Silent Mutation | TTC,TTT | F,F 138 | XP_016870506.1 | |
XM_017015018.1 | 762 | Silent Mutation | TTC,TTT | F,F 138 | XP_016870507.1 |