Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTTAGGTTAATAACTGCCAGGCA[C/G]CCGGAGAGGAGAGCAGGAAGACACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 121013 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GJA5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GJA5 - gap junction protein alpha 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005266.6 | 4812 | UTR 3 | NP_005257.2 | |||
NM_181703.3 | 4812 | UTR 3 | NP_859054.1 | |||
XM_005272951.4 | 4812 | UTR 3 | XP_005273008.1 | |||
XM_017001044.1 | 4812 | UTR 3 | XP_016856533.1 |