Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGAGTATTTGCGCTCCGGCGGGGC[G/T]ATGATCTGCAAGACAGCGCGTGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 102610 MIM: 607613 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACTA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACTA1 - actin, alpha 1, skeletal muscle | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100.3 | 1101 | Silent Mutation | ATA,ATC | I,I 332 | NP_001091.1 |
NUP133 - nucleoporin 133 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |