Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCAAGTACCCATCTTGGAGGGTAC[A/G]GACCCCACATGAGGGTGAGGCTCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
23 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613147 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR205 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR205 - microRNA 205 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR205HG - MIR205 host gene | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001104548.1 | 532 | Missense Mutation | CAG,CGG | Q,R 50 | NP_001098018.1 | |
XM_017002058.1 | 532 | Missense Mutation | CAG,CGG | Q,R 50 | XP_016857547.1 |