Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATTAAACTTAAGCATTGGAAGA[C/T]AAAATTGAGGAAATCTCCCAGAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600713 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSD11B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HSD11B1 - hydroxysteroid 11-beta dehydrogenase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206741.1 | Intron | NP_001193670.1 | ||||
NM_005525.3 | Intron | NP_005516.1 | ||||
NM_181755.2 | Intron | NP_861420.1 |
LOC101930114 - uncharacterized LOC101930114 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |