Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGATAATTAGCAAATAATAAATGT[A/G]TTTTCTTGGTCTGGAAATGGCATGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
22 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
GCSAML PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GCSAML - germinal center associated signaling and motility like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281834.1 | 1488 | Intron | NP_001268763.1 | |||
NM_001281835.1 | 1488 | Intron | NP_001268764.1 | |||
NM_001281836.1 | 1488 | Intron | NP_001268765.1 | |||
NM_001281837.1 | 1488 | Intron | NP_001268766.1 | |||
NM_001281838.1 | 1488 | Intron | NP_001268767.1 | |||
NM_001281853.1 | 1488 | Intron | NP_001268782.1 | |||
NM_145278.4 | 1488 | Intron | NP_660321.1 | |||
XM_017000399.1 | 1488 | Intron | XP_016855888.1 |
GCSAML-AS1 - GCSAML antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OR2C3 - olfactory receptor family 2 subfamily C member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198074.4 | 1488 | UTR 3 | NP_932340.3 |