Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCCCTCCCCTGTTCTACCATGCC[C/T]GGCATGATGGAGAAAGGGCCCGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
RCOR3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RCOR3 - REST corepressor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136223.1 | 220 | Silent Mutation | CCC,CCT | P,P 2 | NP_001129695.1 | |
NM_001136224.2 | 220 | Silent Mutation | CCC,CCT | P,P 2 | NP_001129696.2 | |
NM_001136225.1 | 220 | Silent Mutation | CCC,CCT | P,P 2 | NP_001129697.1 | |
NM_018254.3 | 220 | Intron | NP_060724.1 | |||
XM_005273191.4 | 220 | Silent Mutation | CCC,CCT | P,P 2 | XP_005273248.1 | |
XM_006711442.2 | 220 | Silent Mutation | CCC,CCT | P,P 2 | XP_006711505.1 | |
XM_011509748.2 | 220 | Silent Mutation | CCC,CCT | P,P 2 | XP_011508050.1 | |
XM_011509749.2 | 220 | Silent Mutation | CCC,CCT | P,P 2 | XP_011508051.1 | |
XM_017001758.1 | 220 | Silent Mutation | CCC,CCT | P,P 2 | XP_016857247.1 | |
XM_017001759.1 | 220 | Silent Mutation | CCC,CCT | P,P 2 | XP_016857248.1 | |
XM_017001760.1 | 220 | Silent Mutation | CCC,CCT | P,P 2 | XP_016857249.1 | |
XM_017001761.1 | 220 | Silent Mutation | CCC,CCT | P,P 2 | XP_016857250.1 | |
XM_017001762.1 | 220 | Silent Mutation | CCC,CCT | P,P 2 | XP_016857251.1 | |
XM_017001763.1 | 220 | Silent Mutation | CCC,CCT | P,P 2 | XP_016857252.1 | |
XM_017001764.1 | 220 | Silent Mutation | CCC,CCT | P,P 2 | XP_016857253.1 | |
XM_017001765.1 | 220 | Silent Mutation | CCC,CCT | P,P 2 | XP_016857254.1 | |
XM_017001766.1 | 220 | UTR 5 | XP_016857255.1 | |||
XM_017001767.1 | 220 | Silent Mutation | CCC,CCT | P,P 2 | XP_016857256.1 |