Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAAATGAATCATAATAAAATAGC[C/T]TCTAAACAGTTTCTAAGCGGGAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601546 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PROX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PROX1 - prospero homeobox 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270616.1 | 156 | Intron | NP_001257545.1 | |||
NM_002763.4 | 156 | UTR 5 | NP_002754.2 | |||
XM_005273194.1 | 156 | Intron | XP_005273251.1 | |||
XM_005273195.4 | 156 | Intron | XP_005273252.1 | |||
XM_011509771.1 | 156 | UTR 5 | XP_011508073.1 | |||
XM_011509772.2 | 156 | Intron | XP_011508074.1 | |||
XM_011509773.2 | 156 | Intron | XP_011508075.1 | |||
XM_017001832.1 | 156 | Intron | XP_016857321.1 | |||
XM_017001833.1 | 156 | Intron | XP_016857322.1 |
PROX1-AS1 - PROX1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |