Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACTCCATCTGGGGCTCTTCATAG[A/G]AAAAGGAAAATGGCAGCCTGGCCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
24 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120830 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C4BPA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C4BPA - complement component 4 binding protein alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000715.3 | 365 | Silent Mutation | AGA,AGG | R,R 13 | NP_000706.1 | |
XM_005273251.1 | 365 | Silent Mutation | AGA,AGG | R,R 13 | XP_005273308.1 | |
XM_005273252.4 | 365 | Silent Mutation | AGA,AGG | R,R 13 | XP_005273309.1 |