Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACAAGGTCTTCTCTTAATATTAG[C/G]CTCCTAAATCCTATTACATACACAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615003 MIM: 610626 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDHD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DDHD2 - DDHD domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164232.1 | Intron | NP_001157704.1 | ||||
NM_001164234.1 | Intron | NP_001157706.1 | ||||
NM_015214.2 | Intron | NP_056029.2 | ||||
XM_005273454.2 | Intron | XP_005273511.1 | ||||
XM_005273455.3 | Intron | XP_005273512.1 | ||||
XM_005273456.3 | Intron | XP_005273513.1 | ||||
XM_011544456.1 | Intron | XP_011542758.1 | ||||
XM_017013252.1 | Intron | XP_016868741.1 | ||||
XM_017013253.1 | Intron | XP_016868742.1 | ||||
XM_017013254.1 | Intron | XP_016868743.1 | ||||
XM_017013255.1 | Intron | XP_016868744.1 | ||||
XM_017013256.1 | Intron | XP_016868745.1 |
PLPP5 - phospholipid phosphatase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102559.1 | Intron | NP_001096029.1 | ||||
NM_001102560.1 | Intron | NP_001096030.1 | ||||
NM_032483.3 | Intron | NP_115872.2 | ||||
XM_005273661.4 | Intron | XP_005273718.2 | ||||
XM_005273663.3 | Intron | XP_005273720.2 | ||||
XM_011544670.2 | Intron | XP_011542972.1 | ||||
XM_011544671.1 | Intron | XP_011542973.1 | ||||
XM_011544672.1 | Intron | XP_011542974.1 | ||||
XM_011544674.2 | Intron | XP_011542976.1 | ||||
XM_011544675.2 | Intron | XP_011542977.1 | ||||
XM_017013905.1 | Intron | XP_016869394.1 | ||||
XM_017013906.1 | Intron | XP_016869395.1 | ||||
XM_017013907.1 | Intron | XP_016869396.1 |