Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTTATGTTATAAGCAAGGTTAAG[G/T]TCAGTGTGGTACAGAACTTTCAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602713 MIM: 610700 MIM: 610081 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAM9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADAM9 - ADAM metallopeptidase domain 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HTRA4 - HtrA serine peptidase 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TM2D2 - TM2 domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024380.1 | 2303 | UTR 3 | NP_001019551.1 | |||
NM_001024381.1 | 2303 | UTR 3 | NP_001019552.1 | |||
NM_031940.3 | 2303 | UTR 3 | NP_114146.3 | |||
NM_078473.2 | 2303 | UTR 3 | NP_510882.1 | |||
XM_005273657.3 | 2303 | UTR 3 | XP_005273714.1 | |||
XM_006716408.3 | 2303 | Intron | XP_006716471.1 | |||
XM_011544667.2 | 2303 | UTR 3 | XP_011542969.1 |