Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAGAGTGTGTGGGGTTCAGGGGT[A/T]GGGTGTCGAGGTGTGATCCTGGGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612641 MIM: 610772 | ||||||||||||||||||||
Literature Links: |
ANK1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANK1 - ankyrin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000037.3 | 5824 | UTR 3 | NP_000028.3 | |||
NM_001142445.1 | 5824 | Missense Mutation | AAC,TAC | N,Y 155 | NP_001135917.1 | |
NM_001142446.1 | 5824 | Missense Mutation | AAC,TAC | N,Y 1896 | NP_001135918.1 | |
NM_020475.2 | 5824 | Missense Mutation | AAC,TAC | N,Y 1855 | NP_065208.2 | |
NM_020476.2 | 5824 | Missense Mutation | AAC,TAC | N,Y 1880 | NP_065209.2 | |
NM_020477.2 | 5824 | Missense Mutation | AAC,TAC | N,Y 1718 | NP_065210.2 | |
NM_020478.4 | 5824 | UTR 3 | NP_065211.2 | |||
NM_020480.4 | 5824 | Missense Mutation | AAC,TAC | N,Y 108 | NP_065213.2 | |
XM_005273476.4 | 5824 | UTR 3 | XP_005273533.1 | |||
XM_011544490.2 | 5824 | Missense Mutation | AAC,TAC | N,Y 1968 | XP_011542792.1 | |
XM_011544491.2 | 5824 | UTR 3 | XP_011542793.1 | |||
XM_011544494.2 | 5824 | Missense Mutation | AAC,TAC | N,Y 1953 | XP_011542796.1 | |
XM_011544495.2 | 5824 | UTR 3 | XP_011542797.1 | |||
XM_011544496.2 | 5824 | Missense Mutation | AAC,TAC | N,Y 1943 | XP_011542798.1 | |
XM_011544500.2 | 5824 | Missense Mutation | AAC,TAC | N,Y 1913 | XP_011542802.1 | |
XM_011544501.2 | 5824 | UTR 3 | XP_011542803.1 | |||
XM_011544502.2 | 5824 | Missense Mutation | AAC,TAC | N,Y 1888 | XP_011542804.1 | |
XM_011544503.2 | 5824 | UTR 3 | XP_011542805.1 | |||
XM_011544504.2 | 5824 | Missense Mutation | AAC,TAC | N,Y 1751 | XP_011542806.1 | |
XM_011544505.2 | 5824 | UTR 3 | XP_011542807.1 | |||
XM_017013319.1 | 5824 | Missense Mutation | AAC,TAC | N,Y 1960 | XP_016868808.1 | |
XM_017013320.1 | 5824 | UTR 3 | XP_016868809.1 | |||
XM_017013321.1 | 5824 | Missense Mutation | AAC,TAC | N,Y 1939 | XP_016868810.1 | |
XM_017013322.1 | 5824 | Missense Mutation | AAC,TAC | N,Y 1936 | XP_016868811.1 | |
XM_017013323.1 | 5824 | Missense Mutation | AAC,TAC | N,Y 1935 | XP_016868812.1 | |
XM_017013324.1 | 5824 | Missense Mutation | AAC,TAC | N,Y 1921 | XP_016868813.1 | |
XM_017013325.1 | 5824 | Missense Mutation | AAC,TAC | N,Y 1907 | XP_016868814.1 | |
XM_017013326.1 | 5824 | UTR 3 | XP_016868815.1 | |||
XM_017013327.1 | 5824 | Missense Mutation | AAC,TAC | N,Y 1806 | XP_016868816.1 | |
XM_017013328.1 | 5824 | Missense Mutation | AAC,TAC | N,Y 1791 | XP_016868817.1 | |
XM_017013329.1 | 5824 | UTR 3 | XP_016868818.1 |
MIR486-1 - microRNA 486-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR486-2 - microRNA 486-2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NKX6-3 - NK6 homeobox 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |